Learn More
Med Vet International #78 Straight Root Tip Dental Elevator, Stainless Steel
Supplier:  Med Vet International DE78
Encompass_Preferred
Manufactured for optimal results and precision. Designed to make extractions as trauma-free as possible by cutting the periodontal ligament while easing the tooth from the socket. The result is less tissue damage that promotes quicker healing and fewer complications. The #78 elevator is used by rocking and cutting rather than by lifting and prying the way a conventional elevator is used. Luxating elevators have thin, sharp blades that are used to cut periodontal ligaments. The luxating elevator is used by rocking and cutting vs. an elevator that lifts and pries. Luxating elevators reduce putting pressure on adjacent teeth. One-piece stainless steel construction -- tip will not separate from handle. Provides strength and durability. Unique handle shape creates less force on tooth, reducing chance of tooth fractures. Premium AISI 420 German stainless steel with superior craftsmanship. Non-slip grip premium quality handle.
Catalog No. 50-222-3907
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.