Learn More
Med Vet International 4mm Winged Dental Elevator, Stainless Steel
Supplier:  Med Vet International WE4
Encompass_Preferred
Winged elevators combine the winged shape and thin sharp cutting edge to aid in extractions, The winged shape of the elevator tip matches the curve of the tooth and the extra leverage aids in extraction.
Catalog No. 50-222-3877
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.