Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELF4 Monoclonal Antibody (OTI4A3), TrueMAB™, OriGene
SDP

Catalog No. p-200015702
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3187 Supplier OriGene Supplier No. TA810093
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5′-WGGA-3′. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.

Specifications

Antigen ELF4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ELF4
Gene Accession No. Q99607
Gene Alias ELFR, MEF
Gene Symbols ELF4
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 83-345 of human ELF4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2000
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.