Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566521 Supplier Invitrogen™ Supplier No. PA566521
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDCHSHSRTSL QSSHLWEFVR DLLLSPEENC GILEWEDREQ GIFRVVKSEA LAKMW Highest antigen sequence identity to the following orthologs - mouse 96%, rat 100%.

ELF5 is a member of an epithelium-specific subclass of the Ets transcritpion factor family. In addition to its role in regulating the later stages of terminal differentiation of keratinocytes, it appears to regulate a number of epithelium-specific genes found in tissues containing glandular epithelium such as salivary gland and prostate. It has very low affinity to DNA due to its negative regulatory domain at the amino terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELF5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELF5
Gene Accession No. Q9UKW6
Gene Alias E74 like ETS transcription factor 5; E74-like factor 5; E74-like factor 5 (ets domain transcription factor); E74-like factor 5 ESE-2a; Elf5; elf-5; Epithelium specific Ets-2; epithelium-restricted ESE-1-related Ets factor; Epithelium-specific Ets transcription factor 2; ESE2; ESE-2; ESE-5; ESE-5.; ets transcription factor; ETS-related transcription factor Elf-5
Gene Symbols ELF5
Host Species Rabbit
Immunogen Recombinant Human ELF5. Recombinant protein control fragment (Product #RP-107188).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2001
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.