Learn More
BioVendor, LLC Anti-OVA-IgG1 Mouse ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC 63607661
Encompass
TypeDirect ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-Heparin, Plasma-CitrateSample Requirements5 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below. Calibration Range 1.88–120 U/ml Limit of Detection 0.094 U/ml SUMMARY: IgG is the most abundant immunoglobulin and is the main antibody in the secondary immune response. IgG1, with the heavy chain 1 and molecular weight of 146kDa, occupies about 2/3 of total IgG. By limiting antigen to OVA (ovalbumin), and by measuring only IgG1, mouse immune response is simplified for elucidation. Shibayagis OVA-IgG1 ELISA Kit is useful for specific measurement of mouse anti-OVA IgG1 antibody. Manufactured by FUJIFILM Wako Shibayagi Corporation, Japan. OTHER NAMES: anti-Ovalbumin.
Catalog No. 50-893-007
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.