Learn More
BioVendor, LLC IGFBP-1 Human ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC E01
Encompass
IGFBP-1 Human ELISA (96 wells). Enzyme immunoassay kit for quantitative measurement of Insulin-Like Growth Factor Binding Protein-1 (IGFBP-1) in human serum, plasma, urine, amniotic fluid, milk, saliva, brochial lavage, sputum, cell culture supernatant, and other bodily fluids. QC controls are included with the kit. SAMPLE REQUIREMENT: 20 l/well. CALIBRATION RANGE: 08 ng/ml. ASSAY RANGE: 0 128 ng/ml. DETECTION LIMIT: 0.02 ng/ml. SUMMARY: IGFBP1 is one of six proteins that specifically bind IGF-I and IGF-II in body fluids and tissues. IGFBP1 is particularly important in the study of human female reproductive physiology, energy metabolism, and diabetes. TOPICS: energy metabolism, diabetology, pregnancy. OTHER NAMES: AFBP, IBP1, IBP-1, placental protein 12, PP12, hIGFBP-1. MANUFACTURED BY: Mediagnost GmbH, Germany.
Catalog No. 50-894-003
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.