Learn More
Biomatik Corporation Phospho-BIK (Thr33) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51005
Encompass_Preferred
Phospho-BIK (Thr33) Colorimetric Cell-Based ELISA Kit for detecting Phospho-BIK (Thr33) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13323. Gene ID: 638. Target Synonyms: Apoptosis inducer NBK; BIKLK; BIP1; BP4; Bcl-2 interacting killer; NBK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1214