Learn More
Biomatik Corporation Phospho-Telomerase (Ser824) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51371
Encompass_Preferred
Phospho-Telomerase (Ser824) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Telomerase (Ser824) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O14746. Gene ID: 7015. Target Synonyms: EST2; HEST2; TCS1; Telomerase catalytic subunit; Telomerase reverse transcriptase; TERT; TRT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1531