Learn More
Biomatik Corporation Phospho-Met (Tyr1349) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51508
Encompass_Preferred
Phospho-Met (Tyr1349) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Met (Tyr1349) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08581. Gene ID: 4233. Target Synonyms: c-met; Hepatocyte growth factor receptor precursor; HGF receptor; HGF-SF receptor; kinase Met; Met proto- oncogene tyrosine kinase; met proto-oncogene (hepatocyte growth factor receptor). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1644