Learn More
Biomatik Corporation Phospho-KSR (Ser392) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51023
Encompass_Preferred
Phospho-KSR (Ser392) Colorimetric Cell-Based ELISA Kit for detecting Phospho-KSR (Ser392) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8IVT5. Gene ID: 8844. Target Synonyms: CAP kinase; HB; HB protein; Kinase suppressor of RAS-1; ceramide-activated protein kinase; kinase KSR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1229