Learn More
Biomatik Corporation Phospho-ALK (Tyr1604) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51180
Encompass_Preferred
Phospho-ALK (Tyr1604) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ALK (Tyr1604) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UM73. Gene ID: 238. Target Synonyms: ALK tyrosine kinase receptor precursor; Anaplastic lymphoma kinase; CD246; kinase ALK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1362