Learn More
Biomatik Corporation Phospho-Caspase 9 (Tyr153) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51263
Encompass_Preferred
Phospho-Caspase 9 (Tyr153) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Caspase 9 (Tyr153) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P55211. Gene ID: 842. Target Synonyms: APAF-3; Apoptotic protease Mch-6; Apoptotic protease activating factor 3; CASP-9; Caspase-9; Caspase-9 precursor; Caspase9; ICE-LAP6; ICE-like apoptotic protease 6; MCH6; RNCASP9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1435