Learn More
Biomatik Corporation Phospho-CKII-β (Ser209) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51277
Encompass_Preferred
Phospho-CKII-β (Ser209) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CKII-β (Ser209) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P67870. Gene ID: 1460. Target Synonyms: CK II; CK2-beta; CK2N; CSK2B; CSNK2B; Casein kinase II beta chain; G5a; Phosvitin; beta subunit; casein kinase II; kinase CK2-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1447