Learn More
Biomatik Corporation Phospho-MEK2 (Thr394) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51505
Encompass_Preferred
Phospho-MEK2 (Thr394) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MEK2 (Thr394) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P36507. Gene ID: 5606. Target Synonyms: MAP2K2; Dual specificity mitogen-activated protein kinase kinase 2; MAP kinase kinase 2; MAPKK 2; ERK activator kinase 2; MAPK/ERK kinase 2; MEK2; MEK2; MKK2; PRKMK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1642