Learn More
Biomatik Corporation Phospho-Myc (Thr58) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51513
Encompass_Preferred
Phospho-Myc (Thr58) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Myc (Thr58) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01106. Gene ID: 4609. Target Synonyms: Myc proto-oncogene protein; Class E basic helix-loop-helix protein 39; bHLHe39; Proto-oncogene c-Myc; Transcription factor p64; MYC; BHLHE39. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1648