Learn More
Raybiotech Inc Human VEGF-A ELISA Kit
Supplier:  Raybiotech Inc ELHVEGFCL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human VEGF-A.
Catalog No. 50-194-9847
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.