Learn More
Raybiotech Inc Human Phospho-C-Fos (Thr232) and Total C-Fos ELISA Kit.
Supplier:  Raybiotech Inc PELCFOST232T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated C-Fos (Thr232) and Total C-Fos in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human C-Fos phosphorylated at site Threonine-232 as well as total C-Fos.
Catalog No. 50-195-0539
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.