Learn More
Biomatik Corporation Mouse Inhibitory Subunit Of NF Kappa B Zeta (IkBz) ELISA Kit
Supplier:  Biomatik Corporation EKU11990
Encompass_Preferred
Mouse Inhibitory Subunit Of NF Kappa B Zeta (IkBz) ELISA Kit for detecting IkBz in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 34pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EST8. Gene ID: 80859. Target Synonyms: NFKBIZ; INAP; MAIL; IL-1 inducible nuclear ankyrin-repeat protein; Molecule possessing ankyrin repeats induced by lipopolysaccharide; NF-kappa-B inhibitor zeta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3394
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.