Learn More
Biomatik Corporation Human Calcium/Calmodulin Dependent Protein Kinase II Gamma (CAMK2g) ELISA Kit
Supplier:  Biomatik Corporation EKU12145
Encompass_Preferred
Human Calcium/Calmodulin Dependent Protein Kinase II Gamma (CAMK2g) ELISA Kit for detecting CAMK2g in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13555. Gene ID: 818. Target Synonyms: CAMKII; CAMK2G; CAMK; CAMK-II; CAMKG; CaM Kinase; CaM kinase II subunit gamma; CaMK-II subunit gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3433
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.