Learn More
Biomatik Corporation Human Cubilin (CUBN) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12508
Encompass_Preferred
Human Cubilin (CUBN) Anti-antigen ELISA Kit for detecting CUBN in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.21ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: O60494. Gene ID: 8029. Target Synonyms: IFCR; MGA1; gp280; Intrinsic Factor Cobalamin Receptor; 460 kDa receptor; Intestinal intrinsic factor receptor; Intrinsic factor-vitamin B12 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3537
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.