Learn More
Biomatik Corporation Human Dna-Binding Protein Inhibitor Id-1 (Id1) Elisa Kit
Supplier:  Biomatik Corporation EKC33493
Encompass_Preferred
Human Dna-Binding Protein Inhibitor Id-1 (Id1) Elisa Kit for detecting Inhibitor Of Dna Binding 1, Dominant Negative Helix-Loop-Helix Protein in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P41134; ID1. Target Synonyms: bHLHb24; Class B basic helix-loop-helix protein 24; dJ857M17.1.2 (inhibitor of DNA binding 1; dominant negative helix-loop-helix protein); DNA binding protein inhibitor ID 1; DNA binding protein inhibitor ID1; DNA-binding protein inhibitor ID-1; Dominant negative helix loop helix protein; ID 1; ID; ID1; ID1_HUMAN; Inhibitor of Differentiation 1; Inhibitor of DNA binding 1; inhibitor of DNA binding 1; dominant negative helix-loop-helix protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3932
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.