Learn More
Biomatik Corporation Mouse Collagen Alpha-1 (Xviii) Chain (Col18A1/Es) Elisa Kit
Supplier:  Biomatik Corporation EKC36604
Encompass_Preferred
Mouse Collagen Alpha-1 (Xviii) Chain (Col18A1/Es) Elisa Kit for detecting Collagen, Type Xviii, Alpha 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P39061; Col18a1. Target Synonyms: Col18a1Collagen alpha-1 (XVIII) chain [Cleaved into: Endostatin; Non-collagenous domain 1; NC1)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4386
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.