Learn More
Biomatik Corporation Rat Phospholipid hydroperoxide glutathione peroxidase, mitochondrial(GPX4) ELISA kit
Supplier:  Biomatik Corporation EKC39543
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 4.68 μU/mL-300 μU/mL. Sensitivity: 1.17 μU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P36970. Target Synonyms: MCSP, PHGPx, snGPx, snPHGPx, glutathione peroxidase 4|phospholipid hydroperoxidase|phospholipid hydroperoxide glutathione peroxidase, mitochondrial|sperm nucleus glutathione peroxidase. Restriction: For Research Use Only.
Catalog No. 50-229-4623
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.