Learn More
Biomatik Corporation Pig insulin-like growth factors 2,IGF-2 ELISA Kit
Supplier:  Biomatik Corporation EKC40229
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Sus scrofa (Pig). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P23695. Target Synonyms: C11orf43, FLJ22066, FLJ44734, INSIGF, pp9974, OTTHUMP00000011018|OTTHUMP00000011157|insulin-like growth factor 2|insulin-like growth factor II|insulin-like growth factor type 2|putative insulin-like. Restriction: For Research Use Only.
Catalog No. 50-229-4715
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.