Learn More
Biomatik Corporation Human Insulin-Like Growth Factors 1 Receptor, Igf-1R Elisa Kit
Supplier:  Biomatik Corporation EKC41060
Encompass_Preferred
Human Insulin-Like Growth Factors 1 Receptor, Igf-1R Elisa Kit for detecting Insulin-Like Growth Factor 1 Receptor in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P08069; IGF1R. Target Synonyms: CD221; CD221 antigen; IGF 1 receptor; IGF 1R; IGF I receptor; IGF-I receptor; Igf1r; IGF1R_HUMAN; IGFIR; IGFIRC; IGFR; Insulin like growth factor 1 receptor; Insulin like growth factor 1 receptor precursor; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; MGC142170; MGC142172; MGC18216; Soluble IGF1R variant 1; Soluble IGF1R variant 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4829
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.