Learn More
Biomatik Corporation Human Leukocyte Immunoglobulin-Like Receptor Subfamily A Member 3 (Lilra3) Elisa Kit
Supplier:  Biomatik Corporation EKC41757
Encompass_Preferred
Human Leukocyte Immunoglobulin-Like Receptor Subfamily A Member 3 (Lilra3) Elisa Kit for detecting Leukocyte Immunoglobulin-Like Receptor, Subfamily A (Without Tm Domain), Member 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8N6C8; LILRA3. Target Synonyms: CD 85E; CD85 antigen-like family member E; CD85e; CD85e antigen; e3; HM 31; HM 43; HM31; HM43; ILT 6; ILT-6; ILT6; Immunoglobulin like transcript 6; Immunoglobulin-like transcript 6; Leucocyte immunoglobulin like receptor; Leukocyte immunoglobulin like receptor 4; Leukocyte immunoglobulin-like receptor 4; Leukocyte immunoglobulin-like receptor subfamily A member 3; LILRA 3; LILRA3; LIR 4; LIR-4; LIR4; LIRA3_HUMAN; Monocyte inhibitory receptor HM43/HM31. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4935
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.