Learn More
Biomatik Corporation Human Interferon Induced Transmembrane Protein 3 (IFITM3) ELISA Kit
Supplier:  Biomatik Corporation EKU12069
Encompass_Preferred
Human Interferon Induced Transmembrane Protein 3 (IFITM3) ELISA Kit for detecting IFITM3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01628. Gene ID: 10410. Target Synonyms: 1-8U; IP15; DSPA2b; Dispanin subfamily A member 2b; Interferon-inducible protein 1-8U. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3413
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.