Learn More
Biomatik Corporation Human 25-Hydroxyvitamin D-1 Alpha Hydroxylase, Mitochondrial (Cyp27B1) Elisa Kit
Supplier:  Biomatik Corporation EKC32393
Encompass_Preferred
Human 25-Hydroxyvitamin D-1 Alpha Hydroxylase, Mitochondrial (Cyp27B1) Elisa Kit for detecting Cytochrome P450, Family 27, Subfamily B, Polypeptide 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O15528; CYP27B1. Target Synonyms: 1alpha (OH)ase; 25-hydroxyvitamin D (3) 1-alpha-hydroxylase; 25-hydroxyvitamin D-1 alpha hydroxylase; 25-OHD-1 alpha-hydroxylase; Calcidiol 1-monooxygenase; CP27B_HUMAN; CP2B; CYP1; CYP1ALPHA; CYP27B; Cyp27b1; Cytochrome p450 27B1; Cytochrome p450 27B13; Cytochrome P450 family 27 subfamily B polypeptide 1; Cytochrome P450 subfamily XXVIIB (25-hydroxyvitamin D-1-alpha-hydroxylase) polypeptide 1; Cytochrome P450 subfamily XXVIIB polypeptide 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3753
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.