Learn More
Biomatik Corporation Human Glutamate Carboxypeptidase 2 (Folh1) Elisa Kit
Supplier:  Biomatik Corporation EKC33838
Encompass_Preferred
Human Glutamate Carboxypeptidase 2 (Folh1) Elisa Kit for detecting Folate Hydrolase (Prostate-Specific Membrane Antigen) 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 4.7-300ng/ml. Sensitivity: 1.2ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q04609; FOLH1. Target Synonyms: Cell growth inhibiting protein 27; Cell growth-inhibiting gene 27 protein; FGCP; Folate hydrolase (prostate-specific membrane antigen) 1; Folate hydrolase 1; Folate hydrolase; Folate hydrolase prostate specific membrane antigen 1; FOLH 1; FOLH; Folh1; FOLH1_HUMAN; Folylpoly gamma glutamate carboxypeptidase; Folylpoly-gamma-glutamate carboxypeptidase; GCP 2; GCP II; GCP2; GCPII; GIG27; Glutamate carboxylase II; Glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; Membrane glutamate carboxypeptidase; mGCP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3994
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.