Learn More
Biomatik Corporation Human Herv-K_7P22.1 Provirus Ancestral Env Polyprotein (Ervk6) Elisa Kit
Supplier:  Biomatik Corporation EKC42473
Encompass_Preferred
Human Herv-K_7P22.1 Provirus Ancestral Env Polyprotein (Ervk6) Elisa Kit for detecting Endogenous Retroviral Sequence K, 6 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q69384; ERVK-6. Target Synonyms: ERVK-6; ERVK6Endogenous retrovirus group K member 6 Env polyprotein; EnvK2 protein; Envelope polyprotein; HERV-K (C7) envelope protein; HERV-K (HML-2.HOM) envelope protein; HERV-K108 envelope protein; HERV-K_7p22.1 provirus ancestral Env polyprotein) [Cleaved into: Surface protein; SU); Transmembrane protein; TM)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5092
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.