Learn More
Biomatik Corporation Pig Nuclear Factor-Kappa B, Nf-Κb Elisa Kit
Supplier:  Biomatik Corporation EKC38294
Encompass_Preferred
Pig Nuclear Factor-Kappa B, Nf-Κb Elisa Kit for detecting Nuclear Factor-Kappa B, Nf-Κb in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: NF-kappa B; Nuclear factor-kappa B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4509
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.