Learn More
Biomatik Corporation Human Lysosomal Protective Protein (Ctsa) Elisa Kit
Supplier:  Biomatik Corporation EKC41493
Encompass_Preferred
Human Lysosomal Protective Protein (Ctsa) Elisa Kit for detecting Cathepsin A in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10619; CTSA. Target Synonyms: beta galactosidase 2; BETA GALACTOSIDASE PROTECTIVE PROTEIN; beta-galactosidase 2; beta-galactosidase protective protein; betagalactosidase 2; Carboxypeptidase C; Carboxypeptidase L; carboxypeptidase Y-like kininase; Cathepsin A; Ctsa; deamidase; EC 3.4.16.5; Glactosialidosis; GLB2; Goldberg Syndrome; GSL; lysosomal carboxypeptidase A; Lysosomal protective protein 20 kDa chain; Lysosomal protective protein; Lysosomal protective protein deficiency; NEURAMINIDASE BETA GALACTOSIDASE EXPRESSION; NGBE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4878
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.