Learn More
Biomatik Corporation Human Zinc Finger Protein Aiolos (Ikzf3) Elisa Kit
Supplier:  Biomatik Corporation EKC36077
Encompass_Preferred
Human Zinc Finger Protein Aiolos (Ikzf3) Elisa Kit for detecting Ikaros Family Zinc Finger 3 (Aiolos) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UKT9; IKZF3. Target Synonyms: AIO; Aiolos; IKAROS family zinc finger 3 (Aiolos); IKAROS family zinc finger 3; Ikaros family zinc finger protein 3; IKZF 3; IKZF3; IKZF3_HUMAN; zinc finger DNA binding protein Aiolos; Zinc finger protein Aiolos; Zinc finger protein subfamily 1A 3 (Aiolos); Zinc finger protein subfamily 1A 3; Zinc finger protein subfamily 1A; member 3; ZNFN1A3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4344
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.