Learn More
Biomatik Corporation Human Pancreatic Secretory Trypsin Inhibitor (Spink1) Elisa Kit
Supplier:  Biomatik Corporation EKC42646
Encompass_Preferred
Human Pancreatic Secretory Trypsin Inhibitor (Spink1) Elisa Kit for detecting Pancreatic Secretory Trypsin Inhibitor(Spink1) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00995; SPINK1. Target Synonyms: ISK1_HUMAN; Pancreatic secretory trypsin inhibitor; Pancreatic serine protease inhibitor Kazal; PCTT; PSTI; Serine protease inhibitor Kazal type 1; Serine protease inhibitor Kazal-type 1; Spink 3; SPINK1; Spink3; TaTI; TCP; Tumor associated trypsin inhibitor; Tumor-associated trypsin inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5155
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.