Learn More
Biomatik Corporation Human Epithelial Neutrophil Activating Peptide 78 (Ena-78/Cxcl5) Elisa Kit
Supplier:  Biomatik Corporation EKC40395
Encompass_Preferred
Human Epithelial Neutrophil Activating Peptide 78 (Ena-78/Cxcl5) Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 5 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.567ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P42830; CXCL5. Target Synonyms: AMCFII; C-X-C motif chemokine 5; C-X-C motif chemokine ligand 5; Chemokine (C X C motif) ligand 5; chemokine (C-X-C motif) ligand 5; Cxcl5; CXCL5_HUMAN; ENA 78; ENA-78 (8-78); ENA-78 (1-78); ENA-78 (9-78); ENA78; Epithelial derived neutrophil activating protein 78; Epithelial-derived neutrophil-activating protein 78; Lipopolysaccharide-induced CXC chemokine; Neutrophil activating peptide ENA 78; Neutrophil activating protein 78; Neutrophil-activating peptide ENA-78; neutrophil-activating protein 78; SCYB5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4751
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.