Learn More
Biomatik Corporation Human Regenerating Islet-derived 1 Alpha/pancreatic Stone Protein/pancreatic Thread Protein (REG1A) ELISA kit
Supplier:  Biomatik Corporation EKC40979
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P05451. Target Synonyms: ICRF, MGC12447, P19, PSP, PSPS, PSPS1, PTP, REG, islet cells regeneration factor|islet of langerhans regenerating protein|lithostathine 1 alpha|pancreatic stone protein, secretory|pancreatic thread. Restriction: For Research Use Only.
Catalog No. 50-229-4815
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.