Learn More
Biomatik Corporation Human Epithelial Cell Adhesion Molecule (Epcam) Elisa Kit
Supplier:  Biomatik Corporation EKC42467
Encompass_Preferred
Human Epithelial Cell Adhesion Molecule (Epcam) Elisa Kit for detecting Epithelial Cell Adhesion Molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q3T0L5. Target Synonyms: EPCAM; TACSTD1Epithelial cell adhesion molecule; Ep-CAM; Tumor-associated calcium signal transducer 1; CD antigen CD326. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5089
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.