Learn More
Biomatik Corporation Human GULP1 (PTB domain-containing engulfment adapter protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60426
Encompass_Preferred
Human GULP1 (PTB domain-containing engulfment adapter protein 1) ELISA Kit for detecting GULP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UBP9. Target Synonyms: GULP1; PTB domain-containing engulfment adapter protein 1; Cell death protein 6 homolog; PTB domain adapter protein CED-6; Protein GULP; CED6; GULP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6476
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.