Learn More
Biomatik Corporation Human UBE3A (Ubiquitin-protein ligase E3A) ELISA Kit
Supplier:  Biomatik Corporation EKF60455
Encompass_Preferred
Human UBE3A (Ubiquitin-protein ligase E3A) ELISA Kit for detecting UBE3A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q05086. Target Synonyms: UBE3A; ANCR; EPVE6AP; HPVE6A; ANCREPVE6AP; AS; CTCL tumor antigen se37-2; E6AP; E6-AP; E6AP ubiquitin-protein ligase; HPVE6A; Human papilloma virus E6-associated protein; Human papillomavirus E6-associated protein; Oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54; ubiquitin protein ligase E3A; ubiquitin-protein ligase E3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6493
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.