Learn More
Biomatik Corporation APC Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50036
Encompass_Preferred
APC Colorimetric Cell-Based ELISA Kit for detecting APC in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P25054. Gene ID: 324. Target Synonyms: Adenomatous polyposis coli protein; adenomatous polyposis of the colon gene; APC; APC protein; DP2.5; mAPC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6784
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.