Learn More
Biomatik Corporation Mouse beta cellulin,BTC ELISA Kit
Supplier:  Biomatik Corporation EKC42813
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 pg/ml -4000 pg/ml. Sensitivity: 15.6 pg/ml. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Not Available. Assay Time: 1-5 hours. UniProt ID: Q05928. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5229
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.