Learn More
Biomatik Corporation Mouse LAMP1 (Lysosomal Associated Membrane Protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58953
Encompass_Preferred
Mouse LAMP1 (Lysosomal Associated Membrane Protein 1) ELISA Kit for detecting LAMP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P11438. Target Synonyms: LAMP1; CD107a; CD107 antigen-like family member A; CD107a; CD107a antigen; LAMP-1; LAMPA; LGP120; lysosomal-associated membrane protein 1; lysosome-associated membrane glycoprotein 1; Lysosome-associated membrane protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6180
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.