Learn More
Biomatik Corporation Rat MAPK (P38 Mitogen-Activated Protein Kinase) ELISA Kit
Supplier:  Biomatik Corporation EKF58440
Encompass_Preferred
Rat MAPK (P38 Mitogen-Activated Protein Kinase) ELISA Kit for detecting MAPK in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P70618. Target Synonyms: MAPK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5778
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.