Learn More
Biomatik Corporation Mouse O3far1 (Omega-3 fatty acid receptor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60567
Encompass_Preferred
Mouse O3far1 (Omega-3 fatty acid receptor 1) ELISA Kit for detecting O3far1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7TMA4. Target Synonyms: O3far1; O3FAR1; Omega-3 fatty acid receptor 1; G-protein coupled receptor PGR4; G-protein coupled receptor GT01; G-protein coupled receptor 120; G-protein coupled receptor 129; GPR120; GPR129. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6590
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.