Learn More
Biomatik Corporation Chicken MT (Melatonin) ELISA Kit
Supplier:  Biomatik Corporation EKF60746
Encompass_Preferred
Chicken MT (Melatonin) ELISA Kit for detecting MT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Melatonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6722
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.