Learn More
Biomatik Corporation Human Serum amyloid A, SAA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35538
Encompass_Preferred
Human Serum Amyloid A, Saa Elisa Kit for detecting Serum Amyloid A1 in serum, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 156-10000ng/ml. Sensitivity: 39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P0DJI8; SAA1. Target Synonyms: SAA1; Serum amyloid A-1 protein; SAA) [Cleaved into: Amyloid protein A; Amyloid fibril protein AA); Serum amyloid protein A (2-104); Serum amyloid protein A (3-104); Serum amyloid protein A (2-103); Serum amyloid protein A (2-102); Serum amyloid protein A (4-101)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8265
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.