Learn More
Biomatik Corporation Human Soluble Elastin Fragments (sELAF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35602
Encompass_Preferred
Human Soluble Elastin Fragments (Selaf) Elisa Kit for detecting Soluble Elastin Fragments (Selaf) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8275
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.