Learn More
Biomatik Corporation Human Superoxide dismutase [Mn], mitochondrial (SOD2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35684
Encompass_Preferred
Human Superoxide Dismutase [Mn], Mitochondrial (Sod2) Elisa Kit for detecting Superoxide Dismutase 2, Mitochondrial in serum, plasma, cell culture supernates, cell lysates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04179; SOD2. Target Synonyms: Indophenoloxidase B; IPO B; IPOB; Manganese containing superoxide dismutase; Manganese SOD; Manganese superoxide dismutase; Mangano superoxide dismutase; Mn SOD; Mn superoxide dismutase; MNSOD; MVCD6; SOD 2; SOD2; SODM_HUMAN; Superoxide dismutase [Mn] mitochondrial; Superoxide dismutase [Mn]; mitochondrial; Superoxide dismutase 2 mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.