Learn More
Biomatik Corporation Human tight junction protein 1 (ZO1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35792
Encompass_Preferred
Human Tight Junction Protein 1 (Zo1) Elisa Kit for detecting Tight Junction Protein 1 (Zona Occludens 1) in serum, plasma, cell culture supernates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q07157; TJP1. Target Synonyms: Tight junction protein 1; Tight junction protein ZO-1; Tight junction protein ZO1; TJP1; zo-1; Zo1; ZO1_HUMAN; Zona occludens 1; Zona occludens 1 protein; Zona occludens protein 1; Zonula occludens 1 protein; Zonula occludens protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8316
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.