Learn More
Biomatik Corporation Human Wilms tumor protein (WT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36056
Encompass_Preferred
Human Wilms Tumor Protein (Wt1) Elisa Kit for detecting Wilms Tumor 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19544; WT1. Target Synonyms: WIT 2; WT 1; AWT1; FWT1; GUD; NPHS4; WAGR; Wilms tumor 1; Wilms Tumor; Wilms tumor protein; Wilms' tumor gene; Wilms' tumor protein; WIT2; WT; WT1; WT1_HUMAN; WT33. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8359